Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rimbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081388.1 | NP_001074857.1 | 7 | 815 | ||
GenBank mRNA | AK038814.1 | - | 7 | 509 | ||
AK044685.1 | - | 6 | 915 | |||
AK147417.1 | - | 3 | 219 | |||
BC151100.1 | - | 3 | 406 | 9088013 | ||
BC151101.1 | - | 3 | 406 | 9088014 |