Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C87436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243741.1 | NP_001230670.1 | 7 | 2015 | ||
NM_001243742.1 | NP_001230671.1 | 7 | 1581 | |||
NM_146170.4 | NP_666282.3 | 7 | 1522 | |||
GenBank mRNA | AK047127.1 | - | 7 | 1523 | ||
AK077101.1 | - | 1402 | ||||
AK077503.1 | - | 7 | 1902 | |||
AK170524.1 | - | 8 | 1413 | |||
AK171511.1 | - | 8 | 1413 | |||
AK218862.1 | - | 2 | 134 | |||
BC025632.1 | - | 7 | 1489 | 5321669 | ||
BC026926.1 | - | 8 | 1646 | 3586097 | ||
BC031913.1 | - | 7 | 1535 | 5376246 | ||
BU613064.1 | - | 3 | 240 |