Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Spg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153176.4 | NP_694816.3 | 12, 13 | 1679 | ||
GenBank mRNA | AF512565.1 | - | 12, 13 | 1711 | ||
AF547215.1 | - | 12, 13 | 2008 | |||
AK145540.1 | - | 12, 13 | 1680 | |||
AK171647.1 | - | 9, 10 | 2513 | |||
AK186971.1 | - | 1, 2 | 36 | |||
AK188234.1 | - | 1, 2 | 36 | |||
BC024466.1 | - | 6, 7 | 740 | |||
BC024986.1 | - | 10, 11 | 1334 | |||
BC051051.1 | - | 12, 13 | 1613 | |||
BC055488.1 | - | 1524 | ||||
BC096690.1 | - | 5, 6 | 579 | |||
BC138141.1 | - | 12, 13 | 1906 | 8861160 |