Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOX1 SIRNA Assays ›
Gene Symbol:
Nox1
Gene Name:
NADPH oxidase 1
Gene Aliases:
GP91-2, MOX1, NOH-1, NOH1, NOX1a, NOX1alpha, Nox-1
Chromosome Location:
Chr.X: 134086012 - 134137711 on Build GRCm38
UniGene ID:
Mm.233865
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS215270
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nox1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_172203.2
NP_757340.1
6
727
GenBank mRNA
AF539799.1
-
6
581
AK136432.1
-
6
642
AY174116.1
-
3
322
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053683, AF152963.
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
angiogenesis
regulation of systemic arterial blood pressure by renin-angiotensin
superoxide metabolic process
transport
signal transduction
positive regulation of cell proliferation
proton transport
cell migration
extracellular matrix organization
regulation of ion transmembrane transport
superoxide anion generation
hydrogen peroxide metabolic process
positive regulation of MAPK cascade
positive regulation of integrin biosynthetic process
positive regulation of JNK cascade
positive regulation of smooth muscle cell proliferation
intracellular pH elevation
oxidation-reduction process
cellular response to hyperoxia
oxygen metabolic process
cation transmembrane transport
positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
cellular stress response to acidic pH
positive regulation of reactive oxygen species metabolic process
N-ethylmaleimide reductase activity
superoxide-generating NADPH oxidase activity
oxidoreductase activity
voltage-gated proton channel activity
reduced coenzyme F420 dehydrogenase activity
sulfur oxygenase reductase activity
malolactic enzyme activity
NADPH:sulfur oxidoreductase activity
metal ion binding
Rac GTPase binding
epoxyqueuosine reductase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5hctl:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0