Search
Search

Brip1| Gene Symbol: Brip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178309.2 | NP_840094.1 | 7 | 917 | ||
| GenBank mRNA | AK033205.1 | - | 7 | 889 | ||
| AK051878.1 | - | 7 | 918 | |||
| AK145709.1 | - | 7 | 918 | |||
| AK169440.1 | - | 7 | 864 | |||
| AK169823.1 | - | 7 | 876 | |||
| BC094252.1 | - | 7 | 866 | 6401109 | ||