Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FN3KRP SIRNA Assays ›
          Gene Symbol:
          Fn3krp
          Gene Name:
          fructosamine 3 kinase related protein
          Gene Aliases:
          FN3K-RP
          Chromosome Location:
          Chr.11: 121421373 - 121431288 on Build GRCm38
          UniGene ID:
          Mm.117394
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS215488
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fn3krp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_181420.3 NP_852085.2 6 832
          GenBank mRNA AK078838.1 - 2 252
          AY360466.1 - 6 790
          BC032265.1 - 6 778 4015688

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          phosphorylation
          molecular_function
          phosphofructokinase activity
          phosphorylase kinase regulator activity
          cobinamide kinase activity
          phytol kinase activity
          kinase activity
          phosphatidylinositol phosphate kinase activity
          cyclin-dependent protein serine/threonine kinase regulator activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phenol kinase activity
          cyclin-dependent protein kinase activating kinase regulator activity
          inositol tetrakisphosphate 2-kinase activity
          heptose 7-phosphate kinase activity
          aminoglycoside phosphotransferase activity
          phosphatidylinositol 3-kinase activity
          eukaryotic elongation factor-2 kinase regulator activity
          eukaryotic elongation factor-2 kinase activator activity
          LPPG:FO 2-phospho-L-lactate transferase activity
          cytidine kinase activity
          glycerate 2-kinase activity
          (S)-lactate 2-kinase activity
          phosphoserine:homoserine phosphotransferase activity
          L-seryl-tRNA(Sec) kinase activity
          phosphocholine transferase activity
          polynucleotide 5'-hydroxyl-kinase activity
          ATP-dependent polynucleotide kinase activity
          GTP-dependent polynucleotide kinase activity
          farnesol kinase activity
          CTP:2-trans,-6-trans-farnesol kinase activity
          geraniol kinase activity
          geranylgeraniol kinase activity
          CTP:geranylgeraniol kinase activity
          prenol kinase activity
          1-phosphatidylinositol-5-kinase activity
          1-phosphatidylinositol-3-phosphate 4-kinase activity
          phosphatidylinositol-3,4-bisphosphate 5-kinase activity
          phosphatidylinositol bisphosphate kinase activity
          inositol-3,4,6-trisphosphate 1-kinase activity
          inositol 5-diphosphate pentakisphosphate 5-kinase activity
          inositol diphosphate tetrakisphosphate kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0