Search
Search

Lmln| Gene Symbol: Lmln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172823.2 | NP_766411.1 | 16 | 1799 | ||
| GenBank mRNA | AK030463.1 | - | 16 | 1800 | ||
| BC111899.1 | - | 16 | 1800 | 40062192 | ||
| BC112428.1 | - | 16 | 1800 | 40062196 | ||