Search
Search

Eci2| Gene Symbol: Eci2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110331.1 | NP_001103801.1 | 7 | 834 | ||
| NM_011868.3 | NP_035998.2 | 7 | 1018 | |||
| GenBank mRNA | AF153613.1 | - | 7 | 990 | ||
| AK009478.1 | - | 7 | 818 | |||
| AK170778.1 | - | 7 | 803 | |||
| AK218002.1 | - | 4 | 294 | |||
| BC001983.1 | - | 7 | 728 | 3591615 | ||
| BQ176032.1 | - | 4 | 605 | |||