Search
Search

Nars2| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153591.4 | NP_705819.3 | 2 | 497 | ||
| GenBank mRNA | AK046799.1 | - | 2 | 498 | ||
| AK046801.1 | - | 2 | 498 | |||
| AK047091.1 | - | 2 | 442 | |||
| AK050718.1 | - | 2 | 436 | |||
| AK138260.1 | - | 2 | 477 | |||
| AK164179.1 | - | 2 | 466 | |||
| BB645862.1 | - | 2 | 498 | |||