Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081397.1 | NP_001074866.1 | 32 | 4413 | ||
GenBank mRNA | AB429291.1 | - | 29 | 3919 | ||
AK034257.1 | - | 9 | 1285 | |||
AK220356.1 | - | 13 | 1845 | |||
BC082311.1 | - | 32 | 4279 | |||
BC151049.1 | - | 32 | 4367 | 9087962 | ||
BC151051.1 | - | 32 | 4368 | 9087964 | ||
BC157966.1 | - | 32 | 4368 | 9007290 | ||
BC172122.1 | - | 32 | 4368 | 9054408 |