Search
Search

Eda2r| Gene Symbol: Eda2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161432.1 | NP_001154904.1 | 7 | 996 | ||
| NM_001161433.1 | NP_001154905.1 | 7 | 951 | |||
| NM_175540.5 | NP_780749.3 | 7 | 951 | |||
| GenBank mRNA | AK034909.1 | - | 7 | 915 | ||
| AK049134.1 | - | 7 | 958 | |||
| BC106845.1 | - | 7 | 937 | 40045945 | ||