Search
Search

Eda2r| Gene Symbol: Eda2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161432.1 | NP_001154904.1 | 5, 6 | 603 | ||
| NM_001161433.1 | NP_001154905.1 | 5, 6 | 558 | |||
| NM_175540.5 | NP_780749.3 | 5, 6 | 558 | |||
| GenBank mRNA | AK034909.1 | - | 5, 6 | 522 | ||
| AK049134.1 | - | 5, 6 | 565 | |||
| AK080072.1 | - | 5, 6 | 522 | |||
| BC106845.1 | - | 5, 6 | 544 | 40045945 | ||