Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOD2 SIRNA Assays ›
Gene Symbol:
Nod2
Gene Name:
nucleotide-binding oligomerization domain containing 2
Gene Aliases:
ACUG, BLAU, CD, Card15, F830032C23Rik, IBD1, Nlrc2
Chromosome Location:
Chr.8: 88647280 - 88688474 on Build GRCm38
UniGene ID:
Mm.222633
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS217439
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nod2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_145857.2
NP_665856.2
4
2196
GenBank mRNA
AF520774.1
-
4
2241
AK089390.1
-
4
2160
AY160220.1
-
3
2125
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
innate immune response in mucosa
activation of immune response
cytokine production involved in immune response
cytokine secretion involved in immune response
immune system process
immunoglobulin production involved in immunoglobulin mediated immune response
positive regulation of dendritic cell antigen processing and presentation
negative regulation of T cell mediated immunity
positive regulation of dendritic cell cytokine production
positive regulation of type 2 immune response
negative regulation of inflammatory response to antigenic stimulus
positive regulation of humoral immune response mediated by circulating immunoglobulin
defense response
inflammatory response
positive regulation of antibacterial peptide biosynthetic process
positive regulation of biosynthetic process of antibacterial peptides active against Gram-positive bacteria
signal transduction
positive regulation of cell proliferation
detection of biotic stimulus
detection of bacterium
maintenance of gastrointestinal epithelium
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
response to peptidoglycan
response to muramyl dipeptide
response to lipopolysaccharide
detection of muramyl dipeptide
negative regulation of interferon-gamma production
negative regulation of interleukin-12 production
negative regulation of interleukin-18 production
negative regulation of interleukin-2 production
negative regulation of tumor necrosis factor production
positive regulation of interleukin-1 beta production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-17 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of stress-activated MAPK cascade
negative regulation of toll-like receptor 2 signaling pathway
activation of MAPK activity involved in innate immune response
intracellular signal transduction
defense response to bacterium
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to exogenous dsRNA
positive regulation of MAPK cascade
positive regulation of phosphatidylinositol 3-kinase activity
negative regulation of growth of symbiont in host
innate immune response
positive regulation of innate immune response
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
positive regulation of epithelial cell proliferation
positive regulation of interleukin-1 beta secretion
regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of phagocytosis
defense response to Gram-positive bacterium
positive regulation of B cell activation
positive regulation of NF-kappaB transcription factor activity
protein oligomerization
positive regulation of oxidoreductase activity
positive regulation of nitric-oxide synthase biosynthetic process
positive regulation of prostaglandin-endoperoxide synthase activity
positive regulation of ERK1 and ERK2 cascade
nucleotide-binding oligomerization domain containing 2 signaling pathway
cellular response to peptidoglycan
cellular response to muramyl dipeptide
macrophage inflammatory protein-1 alpha production
regulation of neutrophil chemotaxis
positive regulation of cytokine production involved in inflammatory response
negative regulation of macrophage apoptotic process
positive regulation of prostaglandin-E synthase activity
nucleotide binding
protein binding
ATP binding
endodeoxyribonuclease activity, producing 5'-phosphomonoesters
enzyme binding
protein kinase binding
muramyl dipeptide binding
peptidoglycan binding
CARD domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-zd4m9:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0