Search
Search

Trim61| Gene Symbol: Trim61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177551.1 | NP_001171022.1 | 2 | 1071 | ||
| NM_153110.4 | NP_694750.3 | 3 | 1144 | |||
| GenBank mRNA | AK054544.1 | - | 2 | 957 | ||
| AY063495.1 | - | 2 | 906 | |||
| BC064738.1 | - | 3 | 1022 | 30049322 | ||