Search
Search

Trim61| Gene Symbol: Trim61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177551.1 | NP_001171022.1 | 2 | 980 | ||
| NM_153110.4 | NP_694750.3 | 3 | 1053 | |||
| GenBank mRNA | AK054544.1 | - | 2 | 866 | ||
| AK136037.1 | - | 2 | 895 | |||
| AY063495.1 | - | 2 | 815 | |||
| BC064738.1 | - | 3 | 931 | 30049322 | ||