Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011956.3 | NP_036086.1 | 2, 3 | 194 | ||
GenBank mRNA | AF114169.1 | - | 2, 3 | 172 | ||
AK003816.1 | - | 2, 3 | 192 | |||
AK169896.1 | - | 2, 3 | 153 | |||
BC012635.1 | - | 2, 3 | 195 | 4038123 | ||
BI735707.1 | - | 2, 3 | 194 |