Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vps39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147153.3 | NP_671495.1 | 4 | 343 | ||
NM_178851.3 | NP_849182.1 | 3 | 310 | |||
GenBank mRNA | AB028844.1 | - | 3 | 290 | ||
AF281050.1 | - | 4 | 320 | |||
AF281051.1 | - | 3 | 287 | |||
AK038824.1 | - | 3 | 299 | |||
AK042253.1 | - | 3 | 299 | |||
AK084743.1 | - | 3 | 310 | |||
AK129210.1 | - | 3 | 305 | |||
AK140041.1 | - | 4 | 348 | |||
AK141424.1 | - | 3 | 293 | |||
AK154103.1 | - | 3 | 299 | |||
AK159889.1 | - | 3 | 303 | |||
AK160109.1 | - | 3 | 303 | |||
AK170831.1 | - | 4 | 370 | |||
AK171008.1 | - | 3 | 299 | |||
BB655861.1 | - | 4 | 336 | |||
BC007479.1 | - | 3 | 261 | 3492875 | ||
BY125125.1 | - | 3 | 311 |