Search
Search

Pon3| Gene Symbol: Pon3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173006.1 | NP_766594.1 | 9 | 931 | ||
| GenBank mRNA | AK076090.1 | - | 9 | 1162 | ||
| AK088572.1 | - | 9 | 931 | |||
| BC005714.1 | - | 3 | 291 | |||
| BC099416.1 | - | 9 | 951 | 30847745 | ||
| L76193.1 | - | 9 | 924 | |||