Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175563.5 | NP_780772.1 | 2 | 160 | ||
GenBank mRNA | AK040348.1 | - | 2 | 138 | ||
AK047463.1 | - | 2 | 138 | |||
AK083651.1 | - | 2 | 155 | |||
BC023918.1 | - | 2 | 170 | 5323297 |