Search
Search

Abcg5| Gene Symbol: Abcg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031884.1 | NP_114090.1 | 2 | 217 | ||
| GenBank mRNA | AF312713.2 | - | 2 | 305 | ||
| AK149569.1 | - | 2 | 188 | |||
| AY195872.1 | - | 2 | 305 | |||
| AY195873.1 | - | 2 | 305 | |||
| BC106766.2 | - | 2 | 194 | 40039484 | ||