Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tor1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133673.3 | NP_598434.2 | 5 | 810 | ||
GenBank mRNA | AJ297743.1 | - | 5 | 840 | ||
AK017290.1 | - | 5 | 829 | |||
AK028859.1 | - | 5 | 625 | |||
AK054404.1 | - | 5 | 841 | |||
AK149754.1 | - | 5 | 831 | |||
AK159843.1 | - | 5 | 809 | |||
AK169692.1 | - | 4 | 2594 | |||
AK171756.1 | - | 3 | 1836 | |||
BC018456.1 | - | 5 | 809 | 3582246 |