Search
Search

Dpep2| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301204.1 | NP_001288133.1 | 6 | 890 | ||
| NM_001301205.1 | NP_001288134.1 | 5 | 575 | |||
| NM_176913.4 | NP_795887.2 | 6 | 1014 | |||
| GenBank mRNA | AF488552.1 | - | 6 | 1014 | ||
| AK089165.1 | - | 3 | 2628 | |||
| AK089244.1 | - | 6 | 876 | |||
| AK155099.1 | - | 6 | 842 | |||
| AK155308.1 | - | 6 | 841 | |||
| AK155417.1 | - | 6 | 845 | |||
| AK170094.1 | - | 6 | 842 | |||
| AK170970.1 | - | 6 | 842 | |||
| AK171044.1 | - | 6 | 842 | |||
| AK171122.1 | - | 6 | 842 | |||
| AK171155.1 | - | 5 | 555 | |||
| AK171196.1 | - | 6 | 842 | |||
| AK171268.1 | - | 6 | 842 | |||
| AK171503.1 | - | 6 | 842 | |||
| AK171606.1 | - | 6 | 858 | |||