Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085373.1 | NP_001078842.1 | 11 | 1701 | ||
NM_001085374.1 | NP_001078843.1 | 9 | 1669 | |||
GenBank mRNA | AK054528.1 | - | 9 | 1671 | ||
AK158266.1 | - | 11 | 1702 |