Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nostrin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181547.3 | NP_853525.3 | 14 | 1327 | ||
GenBank mRNA | AJ564264.1 | - | 14 | 1237 | ||
AY280963.1 | - | 14 | 1280 | |||
BC069942.1 | - | 14 | 1304 | 30443942 |