Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Grxcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033426.2 | NP_001028598.1 | 3 | 796 | ||
GenBank mRNA | AK158418.1 | - | 3 | 797 | ||
AK158432.1 | - | 681 | ||||
BC147102.1 | - | 3 | 791 | 9056155 | ||
BC147103.1 | - | 3 | 791 | 9056135 |