Search
Search

Slc41a2| Gene Symbol: Slc41a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177388.3 | NP_796362.1 | 8 | 1461 | ||
| GenBank mRNA | AK038549.1 | - | 8 | 1462 | ||
| BC138146.1 | - | 7 | 1299 | 8861165 | ||
| BC138147.1 | - | 7 | 1299 | 8861166 | ||