Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138628.3 | NP_619534.2 | 3 | 688 | ||
GenBank mRNA | AF422244.1 | - | 3 | 625 | ||
AF422245.1 | - | 3 | 625 | |||
AK052324.1 | - | 3 | 652 | |||
AK084579.1 | - | 3 | 688 | |||
AK084639.1 | - | 3 | 689 | |||
AK085904.1 | - | 3 | 582 | |||
AK137442.1 | - | 3 | 603 | |||
BC141023.1 | - | 3 | 601 | 9055635 |