Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001738.2 | NP_001001738.2 | 2 | 224 | ||
GenBank mRNA | AK160686.1 | - | 2 | 226 | ||
AK165192.1 | - | 2 | 203 | |||
AK171522.1 | - | 2 | 130 | |||
AK171539.1 | - | 2 | 130 | |||
AK171911.1 | - | 2 | 205 | |||
AK220222.1 | - | 2 | 138 | |||
BC063749.1 | - | 2 | 211 | 6813779 |