Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033488.2 | NP_001028660.2 | 8, 9 | 890 | ||
GenBank mRNA | AK133061.1 | - | 8, 9 | 905 | ||
AK156138.1 | - | 9, 10 | 1042 | |||
AK157892.1 | - | 7, 8 | 1717 | |||
AK162620.1 | - | 8, 9 | 889 | |||
AK171660.1 | - | 5, 6 | 553 | |||
AK172423.1 | - | 8, 9 | 891 | |||
BC145584.1 | - | 8, 9 | 903 |