Search
Search

B4galt6| Gene Symbol: B4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019737.2 | NP_062711.1 | 2 | 1129 | ||
| GenBank mRNA | AF097158.1 | - | 2 | 1127 | ||
| AF142674.1 | - | 2 | 337 | |||
| AK137385.1 | - | 2 | 1109 | |||
| AK138247.1 | - | 2 | 1027 | |||
| AK138610.1 | - | 2 | 1111 | |||
| AK144997.1 | - | 2 | 1003 | |||
| AK163956.1 | - | 2 | 1068 | |||
| AK164262.1 | - | 2 | 1068 | |||
| AK164724.1 | - | 2 | 1068 | |||
| BC011149.1 | - | 2 | 352 | 4037384 | ||