Search
Search

Clec4n| Gene Symbol: Clec4n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190320.1 | NP_001177249.1 | 3 | 355 | ||
| NM_001190321.1 | NP_001177250.1 | 3 | 343 | |||
| NM_020001.2 | NP_064385.1 | 4 | 445 | |||
| GenBank mRNA | AF240357.1 | - | 4 | 414 | ||
| AF240358.1 | - | 3 | 312 | |||
| AF240359.1 | - | 4 | 414 | |||
| AK089166.1 | - | 4 | 432 | |||
| AK131697.1 | - | 4 | 432 | |||
| AK154673.1 | - | 3 | 342 | |||
| AK171046.1 | - | 5 | 615 | |||
| BC023008.1 | - | 4 | 414 | 5376247 | ||