Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145937.3 | NP_666049.2 | 2 | 414 | ||
GenBank mRNA | AK151874.1 | - | 2 | 415 | ||
AK159192.1 | - | 2 | 403 | |||
AK160917.1 | - | 2 | 396 | |||
AK161203.1 | - | 2 | 407 | |||
BC026981.1 | - | 2 | 387 | 5365580 |