Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CARM1 SIRNA Assays ›
          Gene Symbol:
          Carm1
          Gene Name:
          coactivator-associated arginine methyltransferase 1
          Gene Aliases:
          Prmt4
          Chromosome Location:
          Chr.9: 21546894 - 21589467 on Build GRCm38
          UniGene ID:
          Mm.178115
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS226869
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Carm1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021531.6 NP_067506.2 14 1675
          NM_153141.1 NP_694781.1 14 1675
          GenBank mRNA AF117887.2 - 14 1585
          AK158242.1 - 14 1640
          AK158757.1 - 14 1515
          BC003964.1 - 9 782
          BC008263.1 - 4 281
          BC026427.1 - 4 241
          BC036974.1 - 14 1659 4935077
          BC071267.1 - 15 1791

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001034088, NM_001030041, AB201116, AB201117, AB201114, and others.

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of growth plate cartilage chondrocyte proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein methylation
          positive regulation of cell proliferation
          pathogenesis
          chromatin modification
          histone methylation
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          intracellular steroid hormone receptor signaling pathway
          intracellular estrogen receptor signaling pathway
          negative regulation of protein binding
          methylation
          regulation of intracellular estrogen receptor signaling pathway
          histone arginine methylation
          histone H3-R2 methylation
          histone H3-R17 methylation
          peptidyl-arginine N-methylation
          positive regulation of fat cell differentiation
          response to cAMP
          endochondral bone morphogenesis
          regulation of mRNA binding
          negative regulation of dendrite development
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          transcription coactivator activity
          protein C-terminal carboxyl O-methyltransferase activity
          tRNA (guanine-N2-)-methyltransferase activity
          protein binding
          methyltransferase activity
          C-methyltransferase activity
          N-methyltransferase activity
          O-methyltransferase activity
          S-methyltransferase activity
          RNA methyltransferase activity
          mRNA methyltransferase activity
          tRNA methyltransferase activity
          protein methyltransferase activity
          2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
          histone-arginine N-methyltransferase activity
          rRNA methyltransferase activity
          rRNA (uridine-2'-O-)-methyltransferase activity
          rRNA (adenine-N6-)-methyltransferase activity
          DNA-methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          rRNA (cytosine-C5-)-methyltransferase activity
          selenocysteine methyltransferase activity
          arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          lysine N-methyltransferase activity
          protein-lysine N-methyltransferase activity
          tRNA (uracil) methyltransferase activity
          tRNA (guanine) methyltransferase activity
          tRNA (adenine) methyltransferase activity
          tRNA (cytosine) methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          tRNA (adenine-N1-)-methyltransferase activity
          rRNA (adenine) methyltransferase activity
          rRNA (cytosine) methyltransferase activity
          rRNA (guanine) methyltransferase activity
          rRNA (uridine) methyltransferase activity
          transferase activity
          protein C-terminal leucine carboxyl O-methyltransferase activity
          1-phenanthrol methyltransferase activity
          protein-arginine N5-methyltransferase activity
          ligand-dependent nuclear receptor transcription coactivator activity
          methylarsonite methyltransferase activity
          dimethylarsinite methyltransferase activity
          4,5-dihydroxybenzo(a)pyrene methyltransferase activity
          1-hydroxypyrene methyltransferase activity
          1-hydroxy-6-methoxypyrene methyltransferase activity
          protein-arginine omega-N asymmetric methyltransferase activity
          histone methyltransferase activity (H3-R17 specific)
          histone methyltransferase activity
          protein homodimerization activity
          demethylmenaquinone methyltransferase activity
          cobalt-precorrin-6B C5-methyltransferase activity
          cobalt-precorrin-7 C15-methyltransferase activity
          cobalt-precorrin-5B C1-methyltransferase activity
          cobalt-precorrin-3 C17-methyltransferase activity
          dimethylamine methyltransferase activity
          hydroxyneurosporene-O-methyltransferase activity
          tRNA (adenine-57, 58-N(1)-) methyltransferase activity
          methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
          trimethylamine methyltransferase activity
          methanol-specific methylcobalamin:coenzyme M methyltransferase activity
          monomethylamine methyltransferase activity
          transcription regulatory region DNA binding
          P-methyltransferase activity
          Se-methyltransferase activity
          2-phytyl-1,4-naphthoquinone methyltransferase activity
          tRNA (uracil-2'-O-)-methyltransferase activity
          tRNA (cytosine-2'-O-)-methyltransferase activity
          phosphomethylethanolamine N-methyltransferase activity
          tRNA (cytosine-3-)-methyltransferase activity
          lysine-acetylated histone binding
          rRNA (cytosine-2'-O-)-methyltransferase activity
          rRNA (cytosine-N4-)-methyltransferase activity
          trihydroxyferuloyl spermidine O-methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cxqm9:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0