Search
Search

Fign| Gene Symbol: Fign | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267846.1 | NP_001254775.1 | 4 | 830 | ||
| NM_001267847.1 | NP_001254776.1 | 4 | 811 | |||
| NM_021716.5 | NP_068362.1 | 3 | 772 | |||
| GenBank mRNA | AF263913.1 | - | 3 | 722 | ||
| AK143787.1 | - | 3 | 747 | |||
| AK163474.1 | - | 3 | 743 | |||
| AK164536.1 | - | 4 | 783 | |||
| BC059266.1 | - | 4 | 830 | 6409940 | ||