Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cntnap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004357.2 | NP_001004357.2 | 24 | 4188 | ||
NM_025771.3 | NP_080047.1 | 4 | 635 | |||
GenBank mRNA | AK013139.1 | - | 4 | 603 | ||
AK017341.1 | - | 4 | 630 | |||
AK129232.1 | - | 24 | 4161 | |||
BC029826.1 | - | 4 | 584 | |||
BC064467.1 | - | 19 | 3079 | |||
BC103799.1 | - | 4 | 542 | |||
BC132074.1 | - | 3 | 447 | |||
BC132076.1 | - | 3 | 447 |