Search
Search

Uqcrc2| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025899.2 | NP_080175.1 | 3 | 247 | ||
| GenBank mRNA | AK003975.1 | - | 3 | 220 | ||
| AK004492.1 | - | 3 | 222 | |||
| AK005151.1 | - | 3 | 231 | |||
| AK007669.1 | - | 2 | 111 | |||
| AK075856.1 | - | 3 | 191 | |||
| AK077583.1 | - | 3 | 196 | |||
| AK088103.1 | - | 3 | 201 | |||
| AK167377.1 | - | 3 | 201 | |||
| BC003423.1 | - | 3 | 219 | 2609295 | ||