Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndufb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026684.2 | NP_080960.1 | 2 | 269 | ||
GenBank mRNA | AK002517.1 | - | 2 | 233 | ||
AK151404.1 | - | 2 | 303 | |||
BC031664.1 | - | 2 | 269 | 4219237 | ||
BC043013.1 | - | 2 | 246 | 5683280 |