Search Thermo Fisher Scientific
Gene Symbol: Acss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080575.2 | NP_542142.1 | 12, 13 | 1796 | ||
GenBank mRNA | AB046742.1 | - | 12, 13 | 1785 | ||
AK088244.1 | - | 12, 13 | 1775 | |||
AK146568.1 | - | 12, 13 | 1797 | |||
AK158119.1 | - | 12, 13 | 1791 | |||
AK172412.1 | - | 12, 13 | 1782 | |||
AK172470.1 | - | 12, 13 | 1771 | |||
AK173275.1 | - | 12, 13 | 1757 | |||
BC004097.1 | - | 11 | ||||
BC030930.1 | - | 8, 9 | 970 | 4911679 | ||
BC092278.1 | - | 12, 13 | 1825 | 3710505 |