Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153527.2 | NP_705755.2 | 8 | 919 | ||
GenBank mRNA | AF419292.2 | - | 8 | 1064 | ||
AK005984.1 | - | 8 | 936 | |||
AK139680.1 | - | 892 | ||||
BC048501.1 | - | 8 | 919 | 6705915 |