Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDUFAF5 SIRNA Assays ›
Gene Symbol:
Ndufaf5
Gene Name:
NADH dehydrogenase (ubiquinone) complex I, assembly factor 5
Gene Aliases:
2310003L22Rik
Chromosome Location:
Chr.2: 140170631 - 140205252 on Build GRCm38
UniGene ID:
Mm.287921
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS229693
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ndufaf5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_027093.4
NP_081369.2
7
715
GenBank mRNA
AK009123.1
-
7
782
BC005630.1
-
7
582
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
metabolic process
methylation
mitochondrial respiratory chain complex I assembly
rRNA (adenine-N6,N6-)-dimethyltransferase activity
molecular_function
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0