Search
Search

Dapk1| Gene Symbol: Dapk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285917.1 | NP_001272846.1 | 10, 11 | 1197 | ||
| NM_029653.3 | NP_083929.2 | 10, 11 | 1197 | |||
| NM_134062.2 | NP_598823.1 | 10, 11 | 1082 | |||
| GenBank mRNA | AK145922.1 | - | 10, 11 | 1083 | ||
| AY245540.2 | - | 10, 11 | 1190 | |||
| AY245541.1 | - | 10, 11 | 1183 | |||
| BC021490.1 | - | 8, 9 | 681 | |||
| BC057317.1 | - | 10, 11 | 1102 | 6821014 | ||
| BC060161.1 | - | 10, 11 | 1102 | 6821014 | ||
| X97048.1 | - | 9, 10 | 1032 | |||
Set Membership: |
|