Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam135b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177819.3 | NP_808487.2 | 15 | 3652 | ||
GenBank mRNA | AK005816.1 | - | 3 | 520 | ||
BC119174.1 | - | 3 | 268 | |||
BC137579.1 | - | 3 | 268 |