Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 3 | 607 | ||
GenBank mRNA | AB055406.1 | - | 3 | 639 | ||
AK018128.1 | - | 3 | 423 | |||
AK036605.1 | - | 3 | 608 | |||
AK042557.1 | - | 3 | 320 | |||
AK047657.1 | - | 3 | 664 | |||
AK086092.1 | - | 3 | 616 | |||
AK138268.1 | - | 3 | 306 | |||
AK160904.1 | - | 3 | 614 | |||
AK161526.1 | - | 3 | 616 | |||
AK162725.1 | - | 3 | 608 | |||
AK163617.1 | - | 3 | 306 | |||
AK165529.1 | - | 3 | 404 | |||
BC026658.1 | - | 3 | 370 | |||
BC116246.1 | - | 3 | 529 | 40093530 | ||
BC116247.1 | - | 3 | 527 | 40093536 |