Search
Search

Rgs12| Gene Symbol: Rgs12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163512.1 | NP_001156984.1 | 4 | 474 | ||
| NM_173402.2 | NP_775578.2 | 7 | 2521 | |||
| GenBank mRNA | AK004813.2 | - | 5 | 427 | ||
| AK014569.1 | - | 4 | 476 | |||
| AK145890.1 | - | 7 | 2469 | |||
| AK161129.1 | - | 4 | 475 | |||
| BC040396.1 | - | 7 | 2518 | 4461061 | ||