Search
Search

Tmprss6| Gene Symbol: Tmprss6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027902.2 | NP_082178.2 | 2, 3 | 454 | ||
| GenBank mRNA | AK004939.2 | - | 2, 3 | 301 | ||
| AY240929.1 | - | 2, 3 | 431 | |||
| BC029645.1 | - | 2, 3 | 431 | 4195486 | ||
| BC057674.1 | - | 2, 3 | 396 | 4162007 | ||
| EU190436.1 | - | 1, 2 | 245 | |||
| EU190437.1 | - | 1, 2 | 245 | |||