Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem55a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028264.4 | NP_082540.1 | 6 | 703 | ||
GenBank mRNA | AK012054.1 | - | 6 | 663 | ||
AK033845.1 | - | 6 | 666 | |||
AK049213.1 | - | 6 | 704 | |||
AX775959.1 | - | 6 | 632 | |||
BC021435.1 | - | 6 | 648 | 5029766 |