Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP13 SIRNA Assays ›
Gene Symbol:
Usp13
Gene Name:
ubiquitin specific peptidase 13 (isopeptidase T-3)
Gene Aliases:
2700071E21Rik, AI848077, ISOT3, IsoT-3
Chromosome Location:
Chr.3: 32817019 - 32938075 on Build GRCm38
UniGene ID:
Mm.316153
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS231684
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Usp13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001013024.2
NP_001013042.1
14
1821
GenBank mRNA
AK135141.1
-
14
1873
BC090999.1
-
14
1821
6754549
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_003940, AK290338, U75362, AK302404, BC016146.
This siRNA also targets the following transcripts in Rat: NM_001107665, XM_008760924.
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
proteolysis
ubiquitin-dependent protein catabolic process
autophagy
cell proliferation
regulation of autophagy
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein stabilization
regulation of proteasomal protein catabolic process
protein K63-linked deubiquitination
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
omega peptidase activity
zinc ion binding
hydrolase activity
ubiquitin protein ligase binding
thiol-dependent ubiquitinyl hydrolase activity
ubiquitin binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System