Search
Search

Ankrd42| Gene Symbol: Ankrd42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028665.4 | NP_082941.2 | 7 | 1831 | ||
| GenBank mRNA | AK016847.1 | - | 2 | 112 | ||
| AK029511.1 | - | 7 | 1833 | |||
| AK029888.1 | - | 7 | 1792 | |||
| AK033599.1 | - | 7 | 1820 | |||
| AK048925.1 | - | 7 | 1725 | |||
| AK133339.1 | - | 1792 | ||||
| BC030885.1 | - | 7 | 1460 | |||
| BC100433.1 | - | 7 | 1867 | |||
| BC145456.1 | - | 7 | 949 | 9054017 | ||
| BC145457.1 | - | 6 | 813 | |||
| BC145458.1 | - | 6 | 813 | |||