Search
Search

Nmnat3| Gene Symbol: Nmnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144533.2 | NP_653116.1 | 2 | 495 | ||
| GenBank mRNA | AK016744.1 | - | 2 | 220 | ||
| AK040783.1 | - | 2 | 476 | |||
| AK041526.1 | - | 2 | 224 | |||
| BC005737.1 | - | 2 | 495 | 3599134 | ||
| BC092086.1 | - | 2 | 204 | 5040019 | ||