Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dmgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028772.3 | NP_083048.1 | 7, 8 | 1180 | ||
GenBank mRNA | AK004755.1 | - | 7, 8 | 1181 | ||
BC024126.1 | - | 3, 4 | 463 | |||
BC089599.1 | - | 7, 8 | 1183 | 30118948 |